3-Fluoro-2-methylcinnamic acid 97%
Dodavatel: Apollo Scientific
PC302300-1G
PC302300-5G
PC302300-25G
APOSPC302300-1GEA
2360
CZK
APOSPC302300-1G
APOSPC302300-5G
APOSPC302300-25G
3-Fluoro-2-methylcinnamic acid 97%
3-Fluoro-2-methylcinnamic acid
Vzorec:
C₁₀H₉FO₂ MW: 180,18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06205416 CAS číslo: 155814-23-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...