Tisk…

3'-Fluoro-2'-(trifluoromethyl)acetophenone 98%

Dodavatel: Apollo Scientific

PC302298-1G PC302298-5G PC302298-10G PC302298-25G
APOSPC302298-1GEA 4370 CZK
APOSPC302298-1G APOSPC302298-5G APOSPC302298-10G APOSPC302298-25G
3'-Fluoro-2'-(trifluoromethyl)acetophenone 98%
3'-Fluoro-2'-(trifluoromethyl)acetophenone
Vzorec: C₉H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD09832249
CAS číslo: 1017777-34-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR