3-Bromo-5-(trifluoromethoxy)benzylamine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC302282-1G
PC302282-5G
APOSPC302282-1GEA
4140
CZK
APOSPC302282-1G
APOSPC302282-5G
3-Bromo-5-(trifluoromethoxy)benzylamine 97%
3-Bromo-5-(trifluoromethoxy)benzylamine
Vzorec:
C₈H₇BrF₃NO Storage Temperature: Ambient |
MDL Number:
MFCD11519380 CAS číslo: 1092461-37-4 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...