3-(3-Fluoro-4-methylphenyl)propionic acid 97%
Dodavatel: Apollo Scientific
PC302264-1G
PC302264-5G
PC302264-10G
APOSPC302264-1GEA
2650
CZK
APOSPC302264-1G
APOSPC302264-5G
APOSPC302264-10G
3-(3-Fluoro-4-methylphenyl)propionic acid 97%
3-(3-Fluoro-4-methylphenyl)propionic acid
Vzorec:
C₁₀H₁₁FO₂ MW: 182,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09025364 CAS číslo: 881189-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...