Tisk…

2-Methyl-3-(trifluoromethyl)benzylamine 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC302261-1G PC302261-5G
APOSPC302261-1GEA 2980 CZK
APOSPC302261-1G APOSPC302261-5G
2-Methyl-3-(trifluoromethyl)benzylamine 98%
2-Methyl-3-(trifluoromethyl)benzylamine
Vzorec: C₉H₁₀F₃N
MW: 189,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06213551
CAS číslo: 771572-43-1
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR