2-Methoxy-6-(trifluoromethyl)benzyl bromide 98%
Dodavatel: Apollo Scientific
PC302259-1G
PC302259-5G
PC302259-10G
APOSPC302259-1GEA
4200
CZK
APOSPC302259-1G
APOSPC302259-5G
APOSPC302259-10G
2-Methoxy-6-(trifluoromethyl)benzyl bromide 98%
2-Methoxy-6-(trifluoromethyl)benzyl bromide
Vzorec:
C₉H₈BrF₃O Storage Temperature: Ambient |
MDL Number:
MFCD09832326 CAS číslo: 1017779-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...