2-Methoxy-4-(trifluoromethoxy)benzonitrile 98%
Dodavatel: Apollo Scientific
PC302249-10G
PC302249-5G
PC302249-1G
APOSPC302249-10GEA
9990
CZK
APOSPC302249-10G
APOSPC302249-5G
APOSPC302249-1G
2-Methoxy-4-(trifluoromethoxy)benzonitrile 98%
2-Methoxy-4-(trifluoromethoxy)benzonitrile
Vzorec:
C₉H₆F₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD04115968 CAS číslo: 886500-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...