Tisk…

2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl alcohol 98%

Dodavatel: Apollo Scientific

Synonyms: 2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl alcohol

PC302244-1G PC302244-5G
APOSPC302244-1GEA 2480 CZK
APOSPC302244-1G APOSPC302244-5G
2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl alcohol 98%
2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl alcohol
Vzorec: C₉H₈F₄O
MW: 208,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687197
CAS číslo: 1323966-16-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR