Tisk…

2'-Fluoro-4',6'-bis(trifluoromethyl)acetophenone 98%

Dodavatel: Apollo Scientific

PC302241-1G PC302241-5G PC302241-10G
APOSPC302241-1GEA 2480 CZK
APOSPC302241-1G APOSPC302241-5G APOSPC302241-10G
2'-Fluoro-4',6'-bis(trifluoromethyl)acetophenone 98%
2'-Fluoro-4',6'-bis(trifluoromethyl)acetophenone
Vzorec: C₁₀H₅F₇O
MW: 274.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258676
CAS číslo: 1017778-58-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR