2-Chloro-5-(pentafluorothio)benzoic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Chloro-5-(pentafluorosulphur)benzoic acid
PC302229-1G
PC302229-5G
PC302229-10G
APOSPC302229-1GEA
9920
CZK
APOSPC302229-1G
APOSPC302229-5G
APOSPC302229-10G
2-Chloro-5-(pentafluorothio)benzoic acid 97%
2-Chloro-5-(pentafluorothio)benzoic acid
Vzorec:
C₇H₄ClF₅O₂S MW: 282,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22988931 CAS číslo: 1431329-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...