Tisk…

2-Chloro-5-(pentafluorothio)benzaldehyde 97%

Dodavatel: Apollo Scientific

PC302228-1G PC302228-5G PC302228-10G
APOSPC302228-1GEA 12010 CZK
APOSPC302228-1G APOSPC302228-5G APOSPC302228-10G
2-Chloro-5-(pentafluorothio)benzaldehyde 97%
2-Chloro-5-(pentafluorothio)benzaldehyde
Vzorec: C₇H₄ClF₅OS
MW: 266,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22988924
CAS číslo: 1431329-76-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR