Tisk…

2,5-Bis(trifluoromethyl)cinnamic acid 97%

Dodavatel: Apollo Scientific

PC302224-1G PC302224-5G
APOSPC302224-1GEA 3580 CZK
APOSPC302224-1G APOSPC302224-5G
2,5-Bis(trifluoromethyl)cinnamic acid 97%
2,5-Bis(trifluoromethyl)cinnamic acid
Vzorec: C₁₁H₆F₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00799486
CAS číslo: 312619-48-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR