Tisk…

2,4-Bis(trifluoromethyl)cinnamic acid 98%

Dodavatel: Apollo Scientific

PC302223-1G PC302223-5G PC302223-10G
APOSPC302223-1GEA 6220 CZK
APOSPC302223-1G APOSPC302223-5G APOSPC302223-10G
2,4-Bis(trifluoromethyl)cinnamic acid 98%
2,4-Bis(trifluoromethyl)cinnamic acid
Vzorec: C₁₁H₆F₆O₂
Storage Temperature: Ambient
MDL Number: MFCD06205287
CAS číslo: 773129-10-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR