Tisk…

4-Methoxy-3-(trifluoromethyl)benzylamine 95%

Dodavatel: Apollo Scientific

PC302218-5G PC302218-1G
APOSPC302218-5GEA 14320 CZK
APOSPC302218-5G APOSPC302218-1G
4-Methoxy-3-(trifluoromethyl)benzylamine 95%
4-Methoxy-3-(trifluoromethyl)benzylamine
Vzorec: C₉H₁₀F₃NO
Storage Temperature: Ambient
MDL Number: MFCD06212984
CAS číslo: 743408-04-0
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR