Tisk…

4-Chloro-3-(trifluoromethoxy)benzylamine 98%

Dodavatel: Apollo Scientific

PC302207-1G PC302207-5G
APOSPC302207-1GEA 4570 CZK
APOSPC302207-1G APOSPC302207-5G
4-Chloro-3-(trifluoromethoxy)benzylamine 98%
4-Chloro-3-(trifluoromethoxy)benzylamine
Vzorec: C₈H₇ClF₃NO
MW: 225,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18393827
CAS číslo: 916210-69-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR