Tisk…

3-Fluoro-6-methoxy-2-(trifluoromethyl)benzaldehyde 97%

Dodavatel: Apollo Scientific

Synonyms: 3-Fluoro-6-methoxy-2-(trifluoromethyl)benzaldehyde

PC302203-1G PC302203-5G PC302203-25G
APOSPC302203-1GEA 2320 CZK
APOSPC302203-1G APOSPC302203-5G APOSPC302203-25G
3-Fluoro-6-methoxy-2-(trifluoromethyl)benzaldehyde 97%
3-Fluoro-6-methoxy-2-(trifluoromethyl)benzaldehyde
Vzorec: C₉H₆F₄O₂
MW: 222,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201030
CAS číslo: 1373921-01-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR