Tisk…

3-Chloro-2-fluoro-6-(trifluoromethyl)benzonitrile 98%

Dodavatel: Apollo Scientific

PC302194-1G PC302194-5G PC302194-10G
APOSPC302194-1GEA 4200 CZK
APOSPC302194-1G APOSPC302194-5G APOSPC302194-10G
3-Chloro-2-fluoro-6-(trifluoromethyl)benzonitrile 98%
3-Chloro-2-fluoro-6-(trifluoromethyl)benzonitrile
Vzorec: C₈H₂ClF₄N
MW: 223.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519382
CAS číslo: 186517-00-0
UN: 3439
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR