Tisk…

2-Methyl-5-(trifluoromethoxy)phenylacetonitrile 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Methyl-5-(trifluoromethoxy)phenylacetonitrile

PC302181-1G PC302181-5G
APOSPC302181-1GEA 3190 CZK
APOSPC302181-1G APOSPC302181-5G
2-Methyl-5-(trifluoromethoxy)phenylacetonitrile 97%
2-Methyl-5-(trifluoromethoxy)phenylacetonitrile
Vzorec: C₁₀H₈F₃NO
MW: 215,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399826
CAS číslo: 1261683-78-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR