Tisk…

2,6-Dichloro-3-(trifluoromethyl)phenylacetic acid 97%

Dodavatel: Apollo Scientific

PC302161-1G PC302161-5G
APOSPC302161-1GEA 3580 CZK
APOSPC302161-1G APOSPC302161-5G
2,6-Dichloro-3-(trifluoromethyl)phenylacetic acid 97%
2,6-Dichloro-3-(trifluoromethyl)phenylacetic acid
Vzorec: C₉H₅Cl₂F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD11519351
CAS číslo: 1092461-11-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR