2,4-Difluorobenzamidoxime 97%
Dodavatel: Apollo Scientific
PC302158-5G
PC302158-10G
PC302158-1G
APOSPC302158-5GEA
2980
CZK
APOSPC302158-5G
APOSPC302158-10G
APOSPC302158-1G
2,4-Difluorobenzamidoxime 97%
2,4-Difluorobenzamidoxime
Vzorec:
C₇H₆F₂N₂O Storage Temperature: Ambient |
CAS číslo:
883022-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...