Tisk…

4-Chloro-3-(trifluoromethyl)-DL-phenylalanine

Dodavatel: Apollo Scientific

PC302146-1G PC302146-5G
APOSPC302146-1GEA 3150 CZK
APOSPC302146-1G APOSPC302146-5G
4-Chloro-3-(trifluoromethyl)-DL-phenylalanine
4-Chloro-3-(trifluoromethyl)-DL-phenylalanine
Vzorec: C₁₀H₉ClF₃NO₂
MW: 267.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09914853
CAS číslo: 63687-03-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR