Tisk…

4-Fluoro-2-methylbenzylamine 98%

Dodavatel: Apollo Scientific

Synonyms: (4-Fluoro-2-methylphenyl)methanamine

PC302111-1G PC302111-5G PC302111-10G
APOSPC302111-1GEA 1080 CZK
APOSPC302111-1G APOSPC302111-5G APOSPC302111-10G
4-Fluoro-2-methylbenzylamine 98%
4-Fluoro-2-methylbenzylamine
Vzorec: C₈H₁₀FN
MW: 139.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701060
CAS číslo: 771574-00-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR