Tisk…

4'-Fluoro-2'-methylacetophenone

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(4-Fluoro-2-methylphenyl)ethanone

PC302110-25G PC302110-5G PC302110-1G
APOSPC302110-25GEA 5230 CZK
APOSPC302110-25G APOSPC302110-5G APOSPC302110-1G
4'-Fluoro-2'-methylacetophenone
4'-Fluoro-2'-methylacetophenone
Vzorec: C₉H₉FO
MW: 152,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00028041
CAS číslo: 446-29-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR