Tisk…

4-Chloro-2-(trifluoromethoxy)benzoyl chloride 97%

Dodavatel: Apollo Scientific

PC302103-1G PC302103-5G PC302103-10G
APOSPC302103-1GEA 1990 CZK
APOSPC302103-1G APOSPC302103-5G APOSPC302103-10G
4-Chloro-2-(trifluoromethoxy)benzoyl chloride 97%
4-Chloro-2-(trifluoromethoxy)benzoyl chloride
Vzorec: C₈H₃Cl₂F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD18393813
CAS číslo: 1261779-42-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR