Tisk…

4,5-Difluoro-2-methoxybenzylamine 98%

Dodavatel: Apollo Scientific

PC302100-1G PC302100-5G
APOSPC302100-1GEA 3310 CZK
APOSPC302100-1G APOSPC302100-5G
4,5-Difluoro-2-methoxybenzylamine 98%
4,5-Difluoro-2-methoxybenzylamine
Vzorec: C₈H₉F₂NO
Storage Temperature: Ambient
MDL Number: MFCD06660190
CAS číslo: 886497-93-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR