3-Methoxy-4-(trifluoromethyl)benzyl bromide 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC302099-1G
PC302099-5G
APOSPC302099-1GEA
2860
CZK
APOSPC302099-1G
APOSPC302099-5G
3-Methoxy-4-(trifluoromethyl)benzyl bromide 98%
3-Methoxy-4-(trifluoromethyl)benzyl bromide
Vzorec:
C₉H₈BrF₃O MW: 269,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18398596 CAS číslo: 853367-87-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...