Tisk…

3-methoxy-4-(trifluoromethyl)benzaldehyde 97%

Dodavatel: Apollo Scientific
Varování

PC302098-1G PC302098-5G PC302098-10G
APOSPC302098-1GEA 2200 CZK
APOSPC302098-1G APOSPC302098-5G APOSPC302098-10G
3-methoxy-4-(trifluoromethyl)benzaldehyde 97%
3-methoxy-4-(trifluoromethyl)benzaldehyde
Vzorec: C₉H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD11110997
CAS číslo: 945003-38-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR