Tisk…

3-Fluoro-2-(trifluoromethyl)benzaldehyde 98%

Dodavatel: Apollo Scientific
Varování

PC302091-1G PC302091-5G PC302091-10G
APOSPC302091-1GEA 992 CZK
APOSPC302091-1G APOSPC302091-5G APOSPC302091-10G
3-Fluoro-2-(trifluoromethyl)benzaldehyde 98%
3-Fluoro-2-(trifluoromethyl)benzaldehyde
Vzorec: C₈H₄F₄O
Storage Temperature: Ambient
MDL Number: MFCD04116354
CAS číslo: 924817-93-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR