3-Chloro-2,6-difluorobenzonitrile 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC302088-5G
PC302088-1G
PC302088-10G
APOSPC302088-5GEA
705
CZK
APOSPC302088-5G
APOSPC302088-1G
APOSPC302088-10G
3-Chloro-2,6-difluorobenzonitrile 98%
3-Chloro-2,6-difluorobenzonitrile
Vzorec:
C₇H₂ClF₂N MW: 173.55 Storage Temperature: Ambient |
MDL Number:
MFCD06660172 CAS číslo: 86225-73-2 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...