Tisk…

3',6'-Dichloro-2'-fluoroacetophenone 98%

Dodavatel: Apollo Scientific

PC302086-1G PC302086-5G PC302086-25G
APOSPC302086-1GEA 2980 CZK
APOSPC302086-1G APOSPC302086-5G APOSPC302086-25G
3',6'-Dichloro-2'-fluoroacetophenone 98%
3',6'-Dichloro-2'-fluoroacetophenone
Vzorec: C₈H₅Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD09025392
CAS číslo: 916420-72-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR