Tisk…

3,5-Difluoro-4-methoxybenzylamine 98%

Dodavatel: Apollo Scientific

PC302085-1G PC302085-5G PC302085-10G
APOSPC302085-1GEA 5130 CZK
APOSPC302085-1G APOSPC302085-5G APOSPC302085-10G
3,5-Difluoro-4-methoxybenzylamine 98%
3,5-Difluoro-4-methoxybenzylamine
Vzorec: C₈H₉F₂NO
MW: 173,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115915
CAS číslo: 105969-16-2
UN: 2922
ADR: 8+6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR