Tisk…

2-Methyl-5-(trifluoromethyl)cinnamic acid 97%

Dodavatel: Apollo Scientific

PC302083-1G PC302083-5G
APOSPC302083-1GEA 2980 CZK
APOSPC302083-1G APOSPC302083-5G
2-Methyl-5-(trifluoromethyl)cinnamic acid 97%
2-Methyl-5-(trifluoromethyl)cinnamic acid
Vzorec: C₁₁H₉F₃O₂
MW: 230,18 g/mol
Bod tání: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD09832288
CAS číslo: 1017778-08-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR