Tisk…

2-Methyl-3-(trifluoromethyl)cinnamic acid 98%

Dodavatel: Apollo Scientific

PC302080-1G PC302080-5G PC302080-10G
APOSPC302080-1GEA 2480 CZK
APOSPC302080-1G APOSPC302080-5G APOSPC302080-10G
2-Methyl-3-(trifluoromethyl)cinnamic acid 98%
2-Methyl-3-(trifluoromethyl)cinnamic acid
Vzorec: C₁₁H₉F₃O₂
MW: 230,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832281
CAS číslo: 1017777-95-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR