Tisk…

5-Chloro-2-(trifluoromethyl)benzonitrile 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC302065-1G PC302065-5G PC302065-10G
APOSPC302065-1GEA 1660 CZK
APOSPC302065-1G APOSPC302065-5G APOSPC302065-10G
5-Chloro-2-(trifluoromethyl)benzonitrile 97%
5-Chloro-2-(trifluoromethyl)benzonitrile
Vzorec: C₈H₃ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD06660352
CAS číslo: 89223-58-5
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR