4-n-Butoxy-2,6-difluorobenzyl alcohol 98%
Dodavatel: Apollo Scientific
Synonyms:
4-butoxy-2,6-difluoro-benzylalcohol
PC302057-1G
PC302057-5G
APOSPC302057-1GEA
1660
CZK
APOSPC302057-1G
APOSPC302057-5G
4-n-Butoxy-2,6-difluorobenzyl alcohol 98%
4-n-Butoxy-2,6-difluorobenzyl alcohol
Vzorec:
C₁₁H₁₄F₂O₂ MW: 216,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22201020 CAS číslo: 438050-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...