3-Chloro-4-(trifluoromethyl)benzonitrile 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Chloro-4-cyanobenzotrifluoride
PC302046-1G
PC302046-5G
PC302046-10G
PC302046-25G
APOSPC302046-1GEA
622
CZK
APOSPC302046-1G
APOSPC302046-5G
APOSPC302046-10G
APOSPC302046-25G
3-Chloro-4-(trifluoromethyl)benzonitrile 98%
3-Chloro-4-(trifluoromethyl)benzonitrile
Vzorec:
C₈H₃ClF₃N MW: 205,57 g/mol Bod tání: 38…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD04972703 CAS číslo: 1092460-79-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...