2-Methoxy-5-(trifluoromethoxy)cinnamic acid 97%
Dodavatel: Apollo Scientific
PC302029-1G
PC302029-5G
PC302029-10G
PC302029-25G
PC302029-100G
APOSPC302029-1GEA
3580
CZK
APOSPC302029-1G
APOSPC302029-5G
APOSPC302029-10G
APOSPC302029-25G
APOSPC302029-100G
2-Methoxy-5-(trifluoromethoxy)cinnamic acid 97%
2-Methoxy-5-(trifluoromethoxy)cinnamic acid
Vzorec:
C₁₁H₉F₃O₄ Storage Temperature: Ambient |
MDL Number:
MFCD09832350 CAS číslo: 1017779-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...