2-[2-Fluoro-5-(trifluoromethyl)phenyl]ethanol 98%
Dodavatel: Apollo Scientific
PC302019-1G
PC302019-5G
PC302019-25G
PC302019-100G
APOSPC302019-1GEA
2940
CZK
APOSPC302019-1G
APOSPC302019-5G
APOSPC302019-25G
APOSPC302019-100G
2-[2-Fluoro-5-(trifluoromethyl)phenyl]ethanol 98%
2-[2-Fluoro-5-(trifluoromethyl)phenyl]ethanol
Vzorec:
C₉H₈F₄O MW: 208,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660358 CAS číslo: 886761-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...