Tisk…

2,6-Dichloro-3-(trifluoromethyl)benzonitrile 98%

Dodavatel: Apollo Scientific

PC302016-1G PC302016-5G PC302016-10G
APOSPC302016-1GEA 2480 CZK
APOSPC302016-1G APOSPC302016-5G APOSPC302016-10G
2,6-Dichloro-3-(trifluoromethyl)benzonitrile 98%
2,6-Dichloro-3-(trifluoromethyl)benzonitrile
Vzorec: C₈H₂Cl₂F₃N
MW: 240,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519347
CAS číslo: 1092461-02-3
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR