2,3-Difluoro-6-methoxyphenol 97%
Dodavatel: Apollo Scientific
Synonyms:
3,4-Difluor-2-hydroxyanisole
PC302010-1G
PC302010-5G
PC302010-10G
APOSPC302010-1GEA
3440
CZK
APOSPC302010-1G
APOSPC302010-5G
APOSPC302010-10G
2,3-Difluoro-6-methoxyphenol 97%
2,3-Difluoro-6-methoxyphenol
Vzorec:
C₇H₆F₂O₂ Bod tání: 44…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD04116024 CAS číslo: 186306-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...