2,3-Difluoro-6-methoxybenzylamine 98%
Dodavatel: Apollo Scientific
PC302009-1G
PC302009-250MG
APOSPC302009-1GEA
20470
CZK
APOSPC302009-1G
APOSPC302009-250MG
2,3-Difluoro-6-methoxybenzylamine 98%
2,3-Difluoro-6-methoxybenzylamine
Vzorec:
C₈H₉F₂NO MW: 173,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04116022 CAS číslo: 886501-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...