Tisk…

Dimethyl octafluoroadipate 98%

Dodavatel: Apollo Scientific

Synonyms: Dimethyl Octafluorohexanedioate, Dimethyl Perfluoroadipate, Octafluoroadipic Acid Dimethyl Ester, Perfluoroadipic Acid Dimethyl Ester, Octafluorohexanedioic Acid Dimethyl Ester

PC3015T-100G PC3015T-25G PC3015T-5G
APOSPC3015T-100GEA 9760 CZK
APOSPC3015T-100G APOSPC3015T-25G APOSPC3015T-5G
Dimethyl octafluoroadipate 98%
Dimethyl octafluoroadipate
Vzorec: C₈H₆F₈O₄
MW: 318,12 g/mol
Bod varu: 120 °C
Hustota: 1,547 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042267
CAS číslo: 3107-98-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR