Tisk…

Dimethyl fluoromalonate 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC3013-25G PC3013-5G PC3013-1G
APOSPC3013-25GEA 744 CZK
APOSPC3013-25G APOSPC3013-5G APOSPC3013-1G
Dimethyl fluoromalonate 97%
Dimethyl fluoromalonate

Active fluoromethylene group, useful reagent for the synthesis of fluoro-heterocycles, e.g Pyrimidines; also can produce fluorinated trisubstituted alkenes, ask for refs.

Vzorec: C₅H₇FO₄
MW: 150,11 g/mol
Bod varu: 111…112 °C/45mmHg
Storage Temperature: Ambient
MDL Number: MFCD00054680
CAS číslo: 344-14-9
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR