Dimethyl dodecafluorosuberate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Dimethyl perfluorosuberate
PC3011-100G
PC3011-25G
PC3011-5G
APOSPC3011-100GEA
16370
CZK
APOSPC3011-100G
APOSPC3011-25G
APOSPC3011-5G
Dimethyl dodecafluorosuberate
Dimethyl dodecafluorosuberate
Vzorec:
C₁₀H₆F₁₂O₄ MW: 418,14 g/mol Bod varu: 152 °C/60mmHg Bod tání: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044088 CAS číslo: 2062-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...