3,4-Dimethylbenzotrifluoride 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC3009T-1G
APOSPC3009T-1GEA
3540
CZK
APOSPC3009T-1G
3,4-Dimethylbenzotrifluoride 97%
3,4-Dimethylbenzotrifluoride
Vzorec:
C₉H₉F₃ Bod varu: 70 °C/10mmHg Storage Temperature: Ambient |
CAS číslo:
78164-31-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...