6-Bromo-3,4-difluoro-2-nitrophenol
Dodavatel: Apollo Scientific
|
Varování
|
PC300970-1G
PC300970-250MG
APOSPC300970-1GEA
9530
CZK
APOSPC300970-1G
APOSPC300970-250MG
6-Bromo-3,4-difluoro-2-nitrophenol
6-Bromo-3,4-difluoro-2-nitrophenol
Vzorec:
C₆H₂BrF₂NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD26099166 CAS číslo: 1644281-87-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...