Tisk…

4-(4-Methyl-1-piperazinyl)-2-(trifluoromethyl)aniline

Dodavatel: Apollo Scientific
Varování

PC300664-1G PC300664-250MG
APOSPC300664-1GEA 5390 CZK
APOSPC300664-1G APOSPC300664-250MG
4-(4-Methyl-1-piperazinyl)-2-(trifluoromethyl)aniline
4-(4-Methyl-1-piperazinyl)-2-(trifluoromethyl)aniline
Vzorec: C₁₂H₁₆F₃N₃
Storage Temperature: Ambient
MDL Number: MFCD09042650
CAS číslo: 863675-89-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR