2-(4-Bromo-1H-pyrazol-1-yl)-4-(trifluoromethyl)pyrimidine
Dodavatel: Apollo Scientific
PC300640-1G
PC300640-10G
PC300640-5G
APOSPC300640-1GEA
6190
CZK
APOSPC300640-1G
APOSPC300640-10G
APOSPC300640-5G
2-(4-Bromo-1H-pyrazol-1-yl)-4-(trifluoromethyl)pyrimidine
2-(4-Bromo-1H-pyrazol-1-yl)-4-(trifluoromethyl)pyrimidine
Vzorec:
C₈H₄BrF₃N₄ MW: 293.05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11505041 CAS číslo: 1215458-83-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...