1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulphonyl chloride
Dodavatel: Apollo Scientific
PC300639-1G
PC300639-5G
APOSPC300639-1GEA
5290
CZK
APOSPC300639-1G
APOSPC300639-5G
1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulphonyl chloride
1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulphonyl chloride
Vzorec:
C₈H₄ClF₃N₄O₂S Storage Temperature: Ambient |
MDL Number:
MFCD11505058 CAS číslo: 1215564-15-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...