Tisk…

4-(4-Fluorophenoxy)benzaldehyde

Dodavatel: Apollo Scientific
Nebezpečí

PC300609-1G PC300609-5G
APOSPC300609-1GEA 2280 CZK
APOSPC300609-1G APOSPC300609-5G
4-(4-Fluorophenoxy)benzaldehyde
4-(4-Fluorophenoxy)benzaldehyde
Vzorec: C₁₃H₉FO₂
MW: 216,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631896
CAS číslo: 137736-06-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR